Learn More
IBA Lifesciences pDSG-IBA Small Mammalian Expression Vector
GREENER_CHOICE
Designed for protein expression for mammalian HEK293E cells (MEXi 293E cells). IBA Lifesciences GmbH Small Mammalian Expression Vector is a small transient expression vector especially developed for the use in combination with the MEXi mammalian expression system.
Brand:  IBA Lifesciences 5-5214-001
 View more versions of this product
Product Code. 15603888
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
pDSG-IBA vectors for MEXi mammalian expression system
  • Small transient expression vector especially developed for the use in combination with the MEXi mammalian expression system.
  • Designed for high-level stable and non-replicative transient expression in most mammalian hosts containing the following elements:
    • Human cytomegalovirus (CMV) immediate-early promoter for high-level expression in a wide range of mammalian cells
    • Episomal replication through Epstein Barr Virus replication origin (oriP) requires chromosomally expressed nuclear antigen encoded by EBNA-1
    • Ampicillin resistance and ColE1 replication origin (pUC) for propagation in E. coliin a high plasmid copy number
Please note that cloning into pDSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pDSG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.
Specifications
Specifications
Constitutive
CMV
Geneticin™ (G-418)
Mammalian
Direct Cloning
Forward
pDSG-IBA45
Ampicillin
Vector
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
His Tag (6x), Strep-tag II
250 ng/μL
5 μg
Vector for recombinant expression in mammalian cells