missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences pDSG-IBA Small Mammalian Expression Vector
GREENER_CHOICE

Product Code. 15693878 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pDSG-IBA102
pDSG-IBA103
pDSG-IBA104
pDSG-IBA105
pDSG-IBA43
pDSG-IBA45
pDSG-IBAwt1
pDSG-IBAwt2
Protein Tag:
His Tag (6x), Strep-tag II
Twin-Strep-tag
Untagged
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15693878

Brand: IBA Lifesciences 55211001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for protein expression for mammalian HEK293E cells (MEXi 293E cells). IBA Lifesciences GmbH Small Mammalian Expression Vector is a small transient expression vector especially developed for the use in combination with the MEXi mammalian expression system.

pDSG-IBA vectors for MEXi mammalian expression system

  • Small transient expression vector especially developed for the use in combination with the MEXi mammalian expression system.
  • Designed for high-level stable and non-replicative transient expression in most mammalian hosts containing the following elements:
    • Human cytomegalovirus (CMV) immediate-early promoter for high-level expression in a wide range of mammalian cells
    • Episomal replication through Epstein Barr Virus replication origin (oriP) requires chromosomally expressed nuclear antigen encoded by EBNA-1
    • Ampicillin resistance and ColE1 replication origin (pUC) for propagation in E. coliin a high plasmid copy number

Please note that cloning into pDSG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pDSG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifications

Constitutive or Inducible System Constitutive
Marker Gene Ampicillin
Promoter CMV
Product Type Vector
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Species Mammalian
Protein Tag His Tag (6x), Strep-tag II
Cloning Method Direct Cloning
Concentration 250 ng/μL
Primer pENTRy-IBA51 Reverse Sequencing
Quantity 5 μg
Vector pDSG-IBA43
For Use With (Application) Vector for recombinant expression in mammalian cells
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.