Learn More
Cole-Parmer™ Small Tough-Spot Labels
Withstand steam-autoclaving, boiling water baths, freezer temperatures, organic solvents, and caustic agents without peeling or falling off. Cole-Parmer™ Small Tough-Spot Labels are made of chemically inert polyester that strongly adheres to all plastics and other materials.
Brand:  Cole-Parmer™ 62207-04
 View more versions of this product
Product Code. 15127082
  • 66.15 € / 1000 rolls
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pre-cut, peel off labels are the ideal size to fit microcentrifuge tubes and other laboratory containers.
  • Withstand temperatures as low as -20°C;-80°C when used with virgin polypropylene tubes.
  • Scuff- and smear-resistant.
Specifications
Specifications
Blue
0.375 mm
–80° when used with virgin polypropylene tubes. Otherwise, –20°C
Polyester
Autoclavable