missing translation for 'onlineSavingsMsg'
Learn More

Labconco Small Tray Dryer, 230V, 50/60Hz

FreeZone Bulk Tray Dryers lyophilize moderate to large bulk sample loads or small batches of vials or serum bottles. FreeZone Console Freeze Dryer and vacuum pump are required.

Brand:  Labconco 780701090

11524.48 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 17940180

  • 12140.00 € / 1 unit
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Lyophilize small to moderate bulk sample loads or batches of vials or serum bottles.
  • Small Tray Dryers have three 10.6 x 11.7 in. shelves that heat up to +60°C. 
  • Designed to fit on either FreeZone Benchtop or Console Freeze Dryers.
  • Lyo-Works™ OS lets the user set up to 30 programs, with up to 16 steps each; record, chart and store data, and receive email alerts.
  • The tray dryer connects to a FreeZone Freeze Dryer that provides a real time display of the tray dryer’s parameters on a large capacitive touchscreen.
  • For use with FreeZone Console Freeze Dryer manufactured after July 2017.
Specifications

Specifications

Small Tray Dryer
60°C
All except North America
3
Stainless Steel
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco Small Tray Dryer, 230V, 50/60Hz > Material: Stainless Steel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.