Learn More
Brady™ Small Universal Condemnation Kit
The Small Universal Condemnation Kit gives you everything you need to lockout gate valves, ball valves, butterfly valves and other valves.
Brand:  Brady™ 806189
Product Code. 11786795
  • 641.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Small adjustable locking kit.
Specifications
Specifications
Small Universal Condemnation Kit
Small adjustable Lockout Kit (FF)
Lockout Tagout
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Small Universal Condemnation Kit > Small Universal Condemnation Kit; Includes: Valve Device Adjustable Steering Wheel, Locking Devices