Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
757.22 € - 1955.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15830182
|
Thermo Scientific™
60115-101 |
SMART Digest Bulk Fractionation Kit, Protein G, Non-Magnetic, Soluble |
1302.74 €
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15840182
|
Thermo Scientific™
60115-102 |
SMART Digest Bulk Fractionation Kit, Protein G, Non-Magnetic, with SOLAμ Collection Plate, Soluble |
1635.00 €
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15850182
|
Thermo Scientific™
60115-103 |
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, with SOLAμ Collection Plate, Soluble |
1830.00 €
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15860182
|
Thermo Scientific™
60115-104 |
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Soluble |
1955.00 €
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15880182
|
Thermo Scientific™
60117-101 |
SMART Digest Bulk Fractionation Kit, Protein G, Magnetic, Standalone |
757.22 €
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
Specifications
Protein G | |
SMART Digest | |
Immunocapture |