Learn More
Thermo Scientific™ SMART Digest™ Chymotrypsin Kits
Obtain fast, simple, and highly reproducible chymotrypsin protein digestion for peptide characterization and quantitation in biopharmaceutical research.
702.56 € - 1360.00 €
Specifications
Capture | None |
---|---|
Cleavage | Peptide bonds of aromatic residues (e.g., tyrosine, phenylalanine, tryptophan) |
Product Line | SMART Digest |
Type | Protein Digestion |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15751717
|
Thermo Scientific™
60109-104 |
SMART Digest Chymotrypsin Kit, with Collection plate |
751.40 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16593432
|
Thermo Scientific™
60109-104-MB-LPH |
SMART Digest Low pH Chymotrypsin Kit, Magnetic Beads, Bulk option for MAM workflows |
817.71 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15698663
|
Thermo Scientific™
60109-104-B |
SMART Digest Chymotrypsin Kit, Bulk Resin option |
702.56 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15608673
|
Thermo Scientific™
60109-104-MB |
SMART Digest Chymotrypsin Kit, Magnetic Bulk Resin option |
793.28 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15761717
|
Thermo Scientific™
60109-105 |
SMART Digest Chymotrypsin Kit, with Filter/Collection plate |
867.72 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15618673
|
Thermo Scientific™
60109-105-B |
SMART Digest Chymotrypsin Kit, Bulk Resin option with Filter/Collection plate |
1185.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15628673
|
Thermo Scientific™
60109-105-MB |
SMART Digest Chymotrypsin Kit, Magnetic Bulk Resin option with Filter/Collection plate |
1200.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15771717
|
Thermo Scientific™
60109-106 |
SMART Digest Chymotrypsin Kit with SOLAμ/collection plate |
1044.84 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15638673
|
Thermo Scientific™
60109-106-B |
SMART Digest Chymotrypsin Kit Bulk Resin option with SOLAμ/collection plate |
1360.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15648673
|
Thermo Scientific™
60109-106-MB |
SMART Digest Chymotrypsin Kit Magnetic Bulk Resin option with SOLAμ/collection plate |
1092.21 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Obtain high-quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Chymotrypsin Kits. A significant advance in sample preparation for biopharmaceutical protein research, these kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. SMART Digest Kits are a significant improvement over in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
Chymotrypsin Protein DigestionCharacterize and quantitate proteins faster, easier, and quicker than ever before using SMART Digest kits. This version of the kit uses chymotrypsin, which cleaves peptide bonds formed by aromatic residues such as tyrosine, phenylalanine, and tryptophan.
Easy and Simple- Significantly increased speed of sample processing
- Simple process
- Easily automate entire protocol for high-throughput processing
- Increased reproducibility over existing protocols
- Higher sensitivity
- Higher data confidence
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digestion can take up to 24 hours.
Cleaner SamplesReduction in the number of chemicals used and the immobilized chymotrypsin design helps mitagate against chemically induced post translational modifications (PTMs) and autolysis. The results are easier to interpret.
Couple with SOLAμ SPE plates for desaltingWhen used with the SMART Digest for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
Specifications
None | |
Peptide bonds of aromatic residues (e.g., tyrosine, phenylalanine, tryptophan) | |
SMART Digest | |
Protein Digestion |