missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ Proteinase K Kits

Product Code. 15618683 Shop All Thermo Scientific Products
Click to view available options
Description:
Bulk Resin option
Bulk Resin option with Filter/Collection plate
Bulk Resin option with SOLAμ/collection plate
Magnetic Bulk Resin option
Magnetic Bulk Resin option with Filter/Collection plate
Magnetic Bulk Resin option with SOLAμ/collection plate
with Collection plate
with Filter/Collection plate
with SOLAμ/collection plate
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15618683

Brand: Thermo Scientific™ 60109109

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well for your biomarker studies.

Obtain high quality analytical results from protein digests by using Thermo Scientific™ SMART Digest™ Proteinase K Kits. A significant advance in sample preparation for biopharmaceutical research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

Proteinase K Protein Digestion

SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses proteinase K, which cleaves the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups.

Easy and Simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized proteinase K design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. The results are easier to interpret.

Couple with SOLAμ SPE Plates for Desalting

When used with the SMART Digest for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

TRUSTED_SUSTAINABILITY

Specifications

Description with SOLAμ/collection plate
Capture None
Cleavage Peptide bond adjacent to carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups
Enzyme Proteinase K
Format SMART Digest Resin Pre-filled into PCR Tubes
For Use With (Application) Peptide Quantitation, Peptide Mapping
Product Line SMART Digest
Type Protein Digestion
Includes SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, SOLAμ SPE Plate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.