Learn More
Cole-Parmer™ Polypropylene Disposable Spatulas, Blue
Reduce cross-contamination with these polypropylene disposable spatulas. Polypropylene Disposable Spatulas are ideal for chemical and biological material transfer, collection, and manipulation. Spatulas feature both a scoop end and a spatula end.
Brand:  Cole-Parmer™ 06265-18
Product Code. 11740579
  • 120.00 € / Pack of 300
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Reduce cross-contamination with these polypropylene disposable spatulas. Polypropylene Disposable Spatulas are ideal for chemical and biological material transfer, collection, and manipulation. Spatulas feature both a scoop end and a spatula end.
  • Disposable polypropylene (PP) spatulas
  • Use as a spatula, stir rod, or policeman
  • Contoured end is shatter resistant even when exposed to liquid nitrogen
  • Flexible enough to fit into many small containers to scrape or collect samples
  • Hollow core reduces heat and cold transfer
  • Can be used with dilute acids and bases
  • Autoclavable
Specifications
Specifications
300 Pk.
Scoop End, Spatula End
Single-use
For chemical and biological material transfer, collection, and manipulation, used with dilute acids and bases, reduce cross-contamination
210 mm
Autoclavable
Blue
Spatula
Polypropylene