Learn More
Thermo Scientific™ Smart-Vue™ Ancillary Parts
Choose from an assortment of ancillary parts including power supplies, cables and mounting hardware.
61.20 €
Specifications
Certifications/Compliance | Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. |
---|---|
For Use With (Application) | Cold Storage |
Description
Choose from an assortment of Thermo Scientific™ Smart-Vue™ Ancillary Parts, including power supplies, cables and mounting hardware for use with Smart-Vue Wireless Monitoring Systems.
Compatible With:
- Refrigerators
- Freezers
- LN2 Storage
- CO2 Incubators
- Clean Rooms
- Storage Areas
- Ovens
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifications
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. | |
Cold Storage |