Learn More
Thermo Scientific™ Smart-Vue Pro Quatro Modules 868Mhz
Increase efficiency with multi-sensor wireless monitoring
Brand:  Thermo Scientific™ SVPHWRMOD024
Product Code. 17501076
  • 767.82 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Smart-Vue Pro Quattro 868Mhz Only. Delivered with two 3.6V lithium batteries, two sensor mounting kit, one mounting holder, one antenna and two 1.5 m/5 ft. flat cable.
  • Bright 2.4 in. color touch screen with multiple views
  • Alarms locally with real -time audio-visual alarms on the modules
  • Audio visual alarms for local notification
  • Communication locally via Bluetooth and to the system via LoRaLow
  • AC power consumption through USB port access
  • Optional replaceable battery power with extended life (minimum of 1 year) allows continuous monitoring
  • Choice of wired and wireless sensors with automatic sensor detection
  • User traceability with customized levels of user access with encrypted password protection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Module
3.6 V lithium battery
Two 3.6 V lithium batteries, two sensor mounting kits, one mounting holder, one antenna, and two 1.5 m/5 ft. flat cables
LCD
2 Years
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products