Learn More
LevGo™ SmartSlicer™ Safety Razor Blades
Say no to dangerous sharps. SmartSlicer™ Safety Razor Blades won't scratch surfaces when cutting agarose gels.
33.50 € - 42.15 €
Specifications
Blade Material Food Grade PET
Product Type Blade
For Use With (Application) Slicing Agarose Gels, Cutting Soft Tissue and Plant Matter
Disposable Single-use
Sterility Non-sterile
View More Specs
Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15326558
View Documents
LevGo™
HS120260
Large
33.50 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
15316558
View Documents
LevGo™
HS120259
Small
42.15 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Cleanly slice agarose gels* right on the light box without scratching the surface.
  • Ideal for cutting soft tissue and plant matter.
  • Adjust the size required by simply cutting with scissors.
  • Available in two sizes and made in the USA from food grade PET.
  • Supplied in packs of 100.
Specifications
Specifications
Food Grade PET
Slicing Agarose Gels, Cutting Soft Tissue and Plant Matter
Non-sterile
Food Grade
No
Blade
Single-use
0.22 mm
100 Pk.