missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMPD1/ASM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14609-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Three transcript variants encoding two different isoforms have been found for this gene.Specifications
SMPD1/ASM | |
Polyclonal | |
Unconjugated | |
SMPD1 | |
ASM, SMPD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20597, 308909, 6609 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17405, Q04519 | |
SMPD1 | |
SMPD1,ASM Fusion Protein Ag6180 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction