missing translation for 'onlineSavingsMsg'
Learn More
3M™ Blue Disposable Coveralls, 4500 Series Product Code.: 13423686

3M™ Blue Disposable Coveralls, 4500 Series

Product Code. 13423686
3X-Large, 76 to 78 in., 194 to 200 cm
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
186 to 194 cm
194 to 200 cm
200 to 206 cm
This item is not returnable. View return policy

Product Code. 13423686

Brand: 3M™ 7000089602

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use for non-hazardous applications. The Blue Disposable Coveralls, 4500 Series offers Category I protection from polypropylene fabric.

CE Simple, Category 1 coverall is designed for domestic DIY use, light-duty industrial cleaning and maintenance applications, or environments where hygiene and cleanliness are important.
  • Excellent economic solution to keep the wearer clean and dry
  • Suitable for non-hazardous applications
  • Breathable and light-duty protection
  • Strong, durable 45gsm polypropylene fabric
  • Storm flap over zip
  • Elasticated hood, cuffs, ankles, and waist
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material Polyethylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Category I
Model 4500B
Size (English) Chest 49 to 52 in.
Size (Metric) Chest 124 to 132 cm
For Use With (Application) Light Industrial Cleaning, Maintenance
Height (English) 76 to 78 in.
Height (Metric) 194 to 200 cm
Show More Show Less
  • 3M Safety Solutions - Protective coveralls overview

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Blue Disposable Coveralls, 4500 Series >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.