missing translation for 'onlineSavingsMsg'
Learn More

Motic™ SMZ171 Series Stereo Zoom Microscope

Rotating the zoom knob gives access to the full zoom range of 6,7:1, resulting in a magnification range of 7.5X-50X with standard WF10X/23mm eyepieces. Motic™ SMZ171 Series Stereo Zoom Microscope makes it simple to drive down from sample overview to tiny structures.

Brand:  Motic™ 1100200600766

1218.04 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15110083

  • 1430.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The Greenough optical system delivers crisp and clear images. Parfocality is ensured while running through the large zoom range of 6,7:1.
  • 5 click-stops within the smooth zoom allow a precise reproduction of magnifications for measuring purposes.
  • Large Field of View is ensured by the standard 10X/23 eyepieces.
  • The large working distance of 110mm gives sufficient space for preparation and repair work, using the stunning depth of view for a convincing 3D upright image.
  • The flexibility of the system allows more complex setups with boom stands, external light sources and an upgrade to a complete documentation station.
Specifications

Specifications

Black/White and frosted glass stage plates and dust cover
7.5X to 50X
23 mm
303 x 239 x 405 mm
2 years electrical / 5 years optical & mechanical parts
SMZ-171-TLED
LED incident and transmitted with independent intensity controls 3w, Transmitted light with tilting reflector (oblique illumination)
1 Each
100/240 V
Achromatic
Zoom 1:6,7
100/240 V
Stereo Microscope with Zoom
Trinocular, 45° inclined, 360° rotating, Fixed optical split 50:50
WF 10X/23 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.