missing translation for 'onlineSavingsMsg'
Learn More
SN 38, Tocris Bioscience™ Product Code.: 18782664

SN 38, Tocris Bioscience™

Product Code. 18782664
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18782664

Brand: Tocris Bioscience 2684/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DNA topoisomerase I inhibitor; antitumor

Active metabolite of CPT-11 (Cat. No. 2688) that inhibits DNA topoisomerase I (IC50 values are 0.74 and 1.9 μM in P388 and Ehrlich cells respectively). Inhibits DNA and RNA synthesis (IC50 values are 0.077 and 1.3 μM respectively) but does not affect protein synthesis. Displays potent antitumor activity against a range of human tumor cell lines (IC50 values are 3.3, 13, 19 and 22 nM for HCT-116, BEL-7402, HL60 and HELA cells respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86639-52-3
Molecular Formula C22H20N2O5
Molecular Weight (g/mol) 392.411
InChI Key FJHBVJOVLFPMQE-QFIPXVFZSA-N
Synonym 7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecinShow MoreShow Less
PubChem CID 104842
ChEBI CHEBI:8988
SMILES CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O

Specifications

Quantity 10 mg
Formula Weight 392.4
Percent Purity >98%
Chemical Name or Material SN 38
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SN 38, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.