missing translation for 'onlineSavingsMsg'
Learn More

sn-Glycero-3-phosphocholine, MedChemExpress

MedChemExpress sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.

POA - 117.45 €

Specifications

Chemical Name or Material sn-Glycero-3-phosphocholine
CAS 28319-77-9
Color White
Purity Grade Notes Research
Molecular Formula C8H20NO6P
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16429867
Encompass_Preferred
View Documents
Medchem Express
HY-17552
5 g
117.45 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16401699
Encompass_Preferred
View Documents
Medchem Express
HY-17552
1 g
55.35 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16417398
Encompass_Preferred
View Documents
Medchem Express
HY-17552
10 g
POA
10g
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
Specifications

Specifications

sn-Glycero-3-phosphocholine
White
C8H20NO6P
H2O : 500 mg/mL (1943.86 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (388.77 mM; Need ultrasonic)
257.22
98.0%
Solid
28319-77-9
Research
Choline Alfoscerate Alpha-GPC L-α-GPC
O=P(OCC[N+](C)(C)C)(OC[C@H](O)CO)[O-]
257.22
Research
Neuroscience-Neurodegeneration
Videos
Safety and Handling

Safety and Handling

H315∣H319∣H335∣H412

missing translation for 'shelfLife' : 4°C, stored under nitrogen, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen, away from moisture)

Recommended Storage : 4°C, stored under nitrogen, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen, away from moisture)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.