missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | SNAI2 |
---|---|
Concentration | 333 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899653
|
Proteintech
12129-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889653
|
Proteintech
12129-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SNAI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SNAI2. This antibody recognizes human, mouse antigen. The SNAI2 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
SNAI2 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Human, Mouse | |
IgG |
333 μg/mL | |
Polyclonal | |
Rabbit | |
6591 | |
Primary |