missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes
GREENER_CHOICE

Improve sample recovery by significantly reducing surface binding with these tubes that do not have additives or coatings.

25.45 € - 56.45 €

Specifications

Cap Style Flat
Color Natural
Material Polypropylene
Graduated Yes
View More Specs

Products 5
Product Code Brand Sterility Capacity (Metric) Price Quantity & Availability  
Product Code Brand Sterility Capacity (Metric) Price Quantity & Availability  
11579914
GREENER_CHOICE
View Documents
Thermo Scientific™
3453
Sterile 2 mL
33.10 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
11535534
GREENER_CHOICE
View Documents
Thermo Scientific™
3434
Non-sterile 2 mL
25.45 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
11569914
GREENER_CHOICE
View Documents
Thermo Scientific™
3451
Sterile 1.5 mL
34.45 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
11525564
GREENER_CHOICE
View Documents
Thermo Scientific™
3446
Non-sterile 0.6 mL
43.75 €
Pack of 1000
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
 
11559914
GREENER_CHOICE
View Documents
Thermo Scientific™
3449
Sterile 0.6 mL
56.45 €
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Improve sample recovery by significantly reducing surface binding with Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes. Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. With no additives or coatings, these tubes — available in 0.6mL, 1.5mL and 2.0mL sizes — offer maximum sample recovery by significantly reducing surface binding.

  • The duckbill cap reduces the pressure required to open the tube with a wider contact surface, yet ensures the cap will remain closed during centrifugation
  • Low retention properties are not achieved by a coating or plastic-embedding processes, ensuring that samples will not be contaminated from these additives
  • Tubes will remain closed during centrifugation yet open easily with one hand as needed
  • Tubes are available in amber to protect your light-sensitive samples
  • Graduation marks on the microcentrifuge tube allow for effective sample reference

Color/Packaging Options

  • A variety of packaging options are available to help with contamination control in the lab
  • For stringent sterility requirements, individually wrapped sterile tubes are available
  • Color options, in addition to natural and amber, are red, blue, green, yellow and violet and a mixed color packaging

Highest Quality

  • Manufactured from virgin polypropylene, these microcentrifuge tubes ensure sample integrity during boiling, freezing and centrifugation
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens

Recommended for:
Daily laboratory usage including centrifugation and short-term sample storage

Specifications

Specifications

Flat
Natural
Polypropylene
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.