missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.
64.56 €
Specifications
Autoclavable | Autoclavable |
---|---|
Capacity (Metric) | 0.5 mL |
Material | Polypropylene |
Temperature Range (English) | -122.8°F to +212°F |
Temperature Range (Metric) | -86°C to +100°C |
Product Code | Brand | Color | Purity or Quality Grade | Packaging | Max. RCF | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Purity or Quality Grade | Packaging | Max. RCF | Price | Quantity & Availability | |||
15615367
![]() |
Eppendorf™
0030123603 |
Clear | Forensic DNA Grade | 500 Tubes (10 Bags x 50 Tubes) | 30,000 x g |
64.56 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
Description
- The hinged Eppendorf Safe-Lock lid prevents unintended opening during incubation and storage.
- g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss in the event of tube breakage and providing enhanced safety when handling hazardous samples.
- Manufactured without the use of slip agents, plasticizers, and biocides, substances known to leach from plastic consumables into the sample and adversely impact bio-assay results.
- Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures.
- Precise lid sealing minimizes evaporation rates during long-term storage.
- The large frosted lid and side surface facilitate easy labeling.
- Ensured functionality in a temperature range from -86 °C to 100 °C.
- Autoclavable when open (121°C, 20 min.) for effective sterilization.
- Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality, ensuring tested purity.
Order Info
Assorted packages include 100 each of natural, blue, green, red and yellow
Specifications
Autoclavable | |
Polypropylene | |
-86°C to +100°C | |
0.02 oz. | |
Cylindrical |
0.5 mL | |
-122.8°F to +212°F | |
Yes | |
Safe-lock Lid | |
Locking Snap-cap Microtube |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube