Learn More
Brady™ Snap-On™ Pipe Cover
Snap-On™ Markers is a fast and easy way to mark rusty, wet or oily pipes. The Snap-On™ Pipe Cover is a transparent PVC cover on which you can apply any of the markers in our ranges. The result is that your self-adhesive marker remains readable and well protected.
Brand:  Brady™ 275240
 View more versions of this product
Product Code. 12779884
  • 18.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Keeps your self-adhesive markers readable and well protected.
Specifications
Specifications
Meet ANSI A13.1 requirements
Brady Snap-On Pipe Markers
Gloss
Pipe Marker
1
1 Ea.
-40°C to +77°C
0.762 mm
228 x 200 x 26 mm
228 mm
200 mm
26 mm
SNAP-ON-1-B817
Transparent
Plastic
Pipe and valve marking
1 Ea.
-40°F to +170°F
0.030 in.
228 x 200 mm
8.98 in.
7.87 in.
1.02 in.