Learn More
Brady™ Snap-On™ Pipe Cover
Snap-On™ Markers is a fast and easy way to mark rusty, wet or oily pipes. The Snap-On™ Pipe Cover is a transparent PVC cover on which you can apply any of the markers in our ranges. The result is that your self-adhesive marker remains readable and well protected.
Brand:  Brady™ 275242
 View more versions of this product
Product Code. 12799884
  • 70.35 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Keeps your self-adhesive markers readable and well protected.
Specifications
Specifications
Meet ANSI A13.1 requirements
Transparent
Plastic
Pipe and valve marking
1 Ea.
-40°F to +170°F
0.030 in.
420 x 420 mm
16.54 in.
16.54 in.
2.05 in.
Brady Snap-On Pipe Markers
Gloss
Pipe Marker
1
1 Ea.
-40°C to +77°C
0.762 mm
420 x 420 x 52 mm
420 mm
420 mm
52 mm