Learn More
Lenz Laborglasintrumente Sockets
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100134
 View more versions of this product
Product Code. 10067720
  • 91.00 € / Pack of 10
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12249 form E and in addition to DIN standard
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
NS 34/35
Duran Glass