Learn More
Lenz Laborglasintrumente Sockets
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100150
 View more versions of this product
Product Code. 10224431
  • 36.57 € / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12249 form E and in addition to DIN standard
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Specifications
NS 50/42
Duran Glass