Learn More
Soda Glass Capillary Tube
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
34.05 € - 46.50 €
Products 3
Product Code Brand Product Type Length (Metric) Tube Price Quantity & Availability  
Product Code Brand Product Type Length (Metric) Tube Price Quantity & Availability  
12807685
View Documents
Samco
G119/02
Capillary Tube 100 mm
34.05 €
Pack of 100
12877675
View Documents
Samco
G119/01
Capillary Tube 75 mm
39.55 €
Pack of 100
12897675
View Documents
Samco
G119/32
Capillary Tube, Both Ends Sealed 100 mm
46.50 €
Pack of 100
Description
Description
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
  • Soda glass capillary tubes
  • Outside diameter 1.8/2.0mm
  • Wall thickness 0.28/0.32mm
  • Packed in tubular cartons of 100