missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan™ General Purpose Specific Gravity Hydrometers Series 50
Shot loaded specific gravity hydrometers for a variety of surface tension ranges.
Brand: Brannan™ 10/911/0
This item is not returnable.
View return policy
Description
- Shot loaded
- Length 190 mm
- Scale subdivisions 0.002
- Accuracy ±0.002 specific gravity
Specifications
1 Ea. | |
0.002 g/mL | |
190 mm |
Specific Gravity Hydrometer | |
UKAS | |
1.20 to 1.25 g/mL |