missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime, ACS Reagent, Honeywell™ Fluka™

Product Code. 15652810
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, granular, +100 mesh particle size

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Physical Form Granular
pH 7.0 - 14.0
CAS 8006-28-8
Material Soda Lime
Quantity 500 g
Molecular Formula CaHNaO2
MDL Number MFCD00134094
Mesh Size +100mesh particle size
UN Number UN1907
Packaging Poly Bottle
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES [OH-].O=[Ca].[Na+]
IUPAC Name sodium;oxocalcium;hydroxide
Molecular Weight (g/mol) 96.074
PubChem CID 66545795
Grade ACS Reagent
Chemical Name or Material Soda Lime
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'