Learn More
Soda lime, ACS, Thermo Scientific Chemicals
61.41 € - 198.99 €
Chemical Identifiers
CAS | 8006-28-8 |
---|---|
Molecular Formula | CaHNaO2 |
Molecular Weight (g/mol) | 96.074 |
MDL Number | MFCD00134094 |
InChI Key | HUAUNKAZQWMVFY-UHFFFAOYSA-M |
PubChem CID | 66545795 |
IUPAC Name | sodium;oxocalcium;hydroxide |
SMILES | [OH-].O=[Ca].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378038
|
Thermo Scientific Alfa Aesar
036596.36 |
500 g |
61.41 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368038
|
Thermo Scientific Alfa Aesar
036596.A4 |
2.5 kg |
198.99 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Soda lime's ability to absorb carbon dioxide makes it valuable in medical and surgical professions. It is also used as powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil CO2 efflux upon absorption. It is also useful for direct measurement of dissolved CO2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8006-28-8 | |
96.074 | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
sodium;oxocalcium;hydroxide |
CaHNaO2 | |
MFCD00134094 | |
66545795 | |
[OH-].O=[Ca].[Na+] |
Specifications
4-8 Mesh | |
Soda Lime | |
MFCD00134094 | |
Soluble in water. | |
[OH-].O=[Ca].[Na+] | |
sodium;oxocalcium;hydroxide | |
66545795 | |
Soda Lime |
8006-28-8 | |
CaHNaO2 | |
4 to 8 | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
Air sensitive; Hygroscopic | |
96.074 | |
ACS Reagent |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H314-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Soda lime
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Calcium hydroxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.