Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
Brand:  Fisherbrand™ 90101010
 View more versions of this product
Product Code. 15832265
  • 18.45 € / Pack of 100
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Withstands moderate warming
  • Light weight
  • Complies with ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 4142
Soda Lime Glass
Yes
100 mm
100 Pack
12 mm