Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Test Tubes
Use Soda Glass Cell Culture Test Tubes to analyze cell culture material that are critical to diagnoses and experimentation.
Brand:  DWK Life Sciences A410013000611
16.11 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11387554
  • 25.40 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Soda lime glass
  • Round bottom
  • X250
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round-bottom
100 mm
250 Pack
13 mm
Test Tube
Soda Lime Glass
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Test Tubes > Rimless; Dimensions: 13 dia. x 100mmH