Learn More
BRAND™ Soda Lime Glass Threaded Bottles
Product Code. 10390092
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2500 mL
Coating:
None
ethylene-acrylate coating
Soda-lime glass bottle features ethylene-acrylate coating that envelops the glass bottle like a protection skin.
  • Soda-lime glass (amber)
  • Screw-cap, polypropene (PP), pouring ring PE-LD
  • Round glass narrow mouth bottles
  • Space-saving square base
  • Available ethylene-acrylate-coated or uncoated
  • The coating envelops the glass bottle like a protection skin
  • Should the bottle break, the splintering hazards are reduced considerably
  • The maximum operating temperature for coated bottles is 80 °C
  • To preserve the coating, do not clean at temperatures exceeding 60 ºC
  • With PP screw cap GL45
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Coating ethylene-acrylate coating
Color Amber
Material Soda Lime Glass