missing translation for 'onlineSavingsMsg'
Learn More
Soda lime, pure, granular, with indicator white to violet, Thermo Scientific Chemicals Product Code.: 10300424

Soda lime, pure, granular, with indicator white to violet, Thermo Scientific Chemicals

Product Code. 10300424
250 g, Glass Bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10300424

Brand: Thermo Scientific Acros 388352500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Gas Adsorbent, Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.074
MDL Number MFCD00134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
PubChem CID 66545795
IUPAC Name sodium;oxocalcium;hydroxide
SMILES [OH-].O=[Ca].[Na+]

Specifications

Color Beige-Gray
Physical Form Granular
Material Soda Lime
Quantity 250 g
Linear Formula CaO/NaOH
Loss on Drying 19.0% max. (10 g, 200°C, 2 Hrs)
Packaging Glass Bottle
Grade Pure
Chemical Name or Material Soda lime

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBS24/038 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number388352500,388350500,10463545,388350010
Lot NumberBS22/051 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number388352500,388350500,10463545,388350010
Lot Number2362202 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number388352500,388350500,10463545,388350010
Lot Number2198468 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number388352500,388350500,10463545,388350010
Lot Number2042170 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number388352500,388350500,10463545,388350010
compliance-icons
Product Identifier
  • Soda lime, granular
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Soda lime, pure, granular, with indicator white to violet, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.