Learn More
Sodium 1-hexanesulfonate monohydrate, 99%, Thermo Scientific Chemicals
Description
Sodium 1-hexanesulfonate monohydrate is an ion pair reagent, which is used for the analysis of small organic compounds, pharmaceutical products and metabolites using high-performance liquid chromatography (HPLC). It is an efficient catalyst for the preparation of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Linear Formula | CH3(CH2)5SO3Na·H2O |
Quantity | 50 g |
Beilstein | 3727014 |
Solubility Information | Slightly soluble in water |
Formula Weight | 206.24 (188.22 Anhydrous) |
Percent Purity | 99% |
Physical Form | Crystalline |
Chemical Name or Material | Sodium 1-Hexanesulfonate Monohydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.