missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-hexanesulfonate monohydrate, 99%, Thermo Scientific Chemicals

Product Code. 11368337
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium 1-hexanesulfonate monohydrate is an ion pair reagent, which is used for the analysis of small organic compounds, pharmaceutical products and metabolites using high-performance liquid chromatography (HPLC). It is an efficient catalyst for the preparation of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207300-91-2
Molecular Formula C6H15NaO4S
Molecular Weight (g/mol) 206.232
MDL Number MFCD00149549
InChI Key CLCGFJYKZGFGSQ-UHFFFAOYSA-M
Synonym 1-hexanesulfonic acid sodium salt monohydrate, sodium hexane-1-sulfonate hydrate, sodium 1-hexanesulfonate monohydrate, n-1-hexanesulfonic acid, sodium 1-hexanesulfonate hydrate, sodium n-hexanesulfonate monohydrate, potassium hexane-1-sulfonate hydrate, sodium hexane-1-sulphonate monohydrate, sodium hexane-1-sulfonate-water 1/1/1, 1-hexanesulfonic acid,sodium salt, hydrate 1:1:1
PubChem CID 23696962
IUPAC Name sodium;hexane-1-sulfonate;hydrate
SMILES CCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Color White
Linear Formula CH3(CH2)5SO3Na·H2O
Quantity 10 g
Beilstein 3727014
Solubility Information Slightly soluble in water
Formula Weight 206.24 (188.22 Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Sodium 1-hexanesulfonate monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.