missing translation for 'onlineSavingsMsg'
Learn More
Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals Product Code.: 11327138

Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals

Product Code. 11327138
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11327138

Brand: Thermo Scientific Alfa Aesar 015764.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium 1-octanesulfonate monohydrate is used as an anionic detergent. It is an ion pairing reagent used in the HPLC analysis of proteins and peptides. Further, it is used as a surface active agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207596-29-0
Molecular Formula C8H19NaO4S
Molecular Weight (g/mol) 234.29
MDL Number MFCD00149551
InChI Key MBURIAHQXJQKRE-UHFFFAOYSA-M
Synonym sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
PubChem CID 23666339
IUPAC Name sodium;octane-1-sulfonate;hydrate
SMILES O.[Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 234.29 (216.28 Anhydrous)
Percent Purity ≥99%
Physical Form Crystalline
Chemical Name or Material Sodium 1-Octanesulfonate Monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM10L038 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog Number015764.18,015764.09
Lot NumberX19K016 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number015764.18,015764.09
Lot NumberW03K016 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number015764.18,015764.09
Lot NumberT26K014 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number015764.18,015764.09
Lot NumberP29K017 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number015764.18,015764.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.