Learn More
Sodium 1-propanethiolate, tech. 85%, Thermo Scientific Chemicals
89.50 € - 347.00 €
Chemical Identifiers
CAS | 6898-84-6 |
---|---|
Molecular Formula | C3H7NaS |
Molecular Weight (g/mol) | 98.139 |
MDL Number | MFCD00043318 |
InChI Key | XVSFHIIADLZQJP-UHFFFAOYSA-M |
Synonym | sodium 1-propanethiolate, sodium propanethiolate, 1-propanethiol sodium salt, sodium propane-1-thiolate, 1-propanethiol, sodium salt, sodium propylsulfanide, propylsulfanylsodium, sodium thiopropoxide, 1-propanethiol, sodium salt 1:1, sodium propyl sulfide |
PubChem CID | 4681725 |
IUPAC Name | sodium;propane-1-thiolate |
SMILES | CCC[S-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15462699
|
Thermo Scientific Alfa Aesar
H66135.06 |
5 g |
89.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472699
|
Thermo Scientific Alfa Aesar
H66135.14 |
25 g |
347.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6898-84-6 | |
98.139 | |
XVSFHIIADLZQJP-UHFFFAOYSA-M | |
4681725 | |
CCC[S-].[Na+] |
C3H7NaS | |
MFCD00043318 | |
sodium 1-propanethiolate, sodium propanethiolate, 1-propanethiol sodium salt, sodium propane-1-thiolate, 1-propanethiol, sodium salt, sodium propylsulfanide, propylsulfanylsodium, sodium thiopropoxide, 1-propanethiol, sodium salt 1:1, sodium propyl sulfide | |
sodium;propane-1-thiolate |
Specifications
6898-84-6 | |
Stench | |
CH3CH2CH2SNa | |
UN3263 | |
Air and moisture sensitive | |
XVSFHIIADLZQJP-UHFFFAOYSA-M | |
sodium;propane-1-thiolate | |
4681725 | |
85% | |
Sodium 1-propanethiolate |
Gray to White | |
C3H7NaS | |
MFCD00043318 | |
3556024 | |
sodium 1-propanethiolate, sodium propanethiolate, 1-propanethiol sodium salt, sodium propane-1-thiolate, 1-propanethiol, sodium salt, sodium propylsulfanide, propylsulfanylsodium, sodium thiopropoxide, 1-propanethiol, sodium salt 1:1, sodium propyl sulfide | |
CCC[S-].[Na+] | |
98.139 | |
98.14 | |
Solid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 230-005-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only