missing translation for 'onlineSavingsMsg'
Learn More
Sodium acetate, 3M aq. soln., pH 5.2, RNAse free, Thermo Scientific Chemicals Product Code.: 15480217

Sodium acetate, 3M aq. soln., pH 5.2, RNAse free, Thermo Scientific Chemicals

Product Code. 15480217
250 mL
Click to view available options
Quantity:
125 mL
250 mL
This item is not returnable. View return policy

Product Code. 15480217

Brand: Thermo Scientific Alfa Aesar J61928.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.

Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, halogens. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agent. .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

pH 5.2
Physical Form Liquid
Quantity 250 mL
Linear Formula NaOOCCH3
Solubility Information Soluble in water (246g/L) at 20°C,and ethanol.
Formula Weight 82.03
Concentration or Composition (by Analyte or Components) 3M aq. soln.
Odor Odorless
Chemical Name or Material Sodium acetate, RNAse free

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ20L506 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog NumberJ61928.AK,J61928.=E,J61928.AE
Lot NumberP27L509 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberJ61928.AK,J61928.AE
Lot NumberX28K530 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberJ61928.AK,J61928.AE
Lot NumberW28K512 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberJ61928.AK,J61928.=E,J61928.AE
Lot NumberW06K532 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberJ61928.AK,J61928.=E,J61928.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium acetate, 3M aq. soln., pH 5.2, RNAse free, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.