missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 99+%, anhydrous, Molecular Biology Reagent, Thermo Scientific Chemicals

Product Code. 15420577
Click to view available options
Quantity:
100 g
500 g

Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

Solubility
Soluble in water (1.19 g/L) at 20°C, and ethanol.

Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324°C
Color White
pH 7.5 to 9
Physical Form Powder
Quantity 500 g
Flash Point >110°C (230°F)
Merck Index 14,8571
Solubility Information Soluble in water (1.19g/L) at 20°C,and ethanol.
IUPAC Name sodium;acetate
Formula Weight 82.03
Percent Purity ≥99%
Sensitivity Hygroscopic
Chemical Name or Material Sodium acetate, Anhydrous, Molecular Biology Reagent
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.