missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Anhydrous, Extra Pure, SLR, Fisher Chemical™
32.17 EUR valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Sodium Acetate Anhydrous, Extra Pure, SLR
Specifications
Melting Point | 324°C |
Color | White |
pH | 7.5 to 9.2 |
Physical Form | Powder/Solid |
Quantity | 1 kg |
IUPAC Name | sodium;acetate |
Formula Weight | 82.03g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'