missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., anhydrous, Honeywell Fluka™

Product Code. 15663810 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
40 kg
6 x 1 kg
Unit Size:
1kg
40kg
500g
5kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15663810

Brand: Honeywell-Fluka 323191KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., anhydrous

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium;acetate
SMILES [Na+].CC([O-])=O

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula CH3COONa
UN Number NONH for all modes of transport
Beilstein 3595639
Flash Point >250°C (>482°F)
Formula Weight 82.03g/mol
Grade ACS/puriss. p.a.
Ignition Point 1112°F
Chemical Name or Material Sodium Acetate, Reag. Ph. Eur.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.