missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, ReagentPlus™, Honeywell™

Product Code. 15688120
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, anhydrous,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Quantity 500 g
Linear Formula CH3COONa
UN Number NONH for all modes of transport
Beilstein 3595639
Flash Point <250°C (>482°F)
IUPAC Name sodium acetate
Formula Weight 82.03g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Ignition Point 1112°F
Chemical Name or Material Sodium acetate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'