missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Alginate, Technical, SLR, Fisher Chemical
Brand: Fisher Chemical S/2160/53
177.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
500g, Plastic powder jar

Specifications
300°C | |
Yellow | |
Powder/Solid | |
(C6H7O7)A(C6H7O7)BNa | |
Plastic powder jar |
9005-38-3 | |
5.5 to 7.5 | |
500 g | |
81310 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Alginate, Technical, SLR, Fisher Chemical > 500g, Plastic powder jar
Spot an opportunity for improvement?Share a Content Correction