missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Arsenite Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
47.90 € - 98.10 €
Chemical Identifiers
CAS | 7784-46-5 |
---|---|
Molecular Formula | AsNaO2 |
Molecular Weight (g/mol) | 129.909 |
MDL Number | 3472 |
InChI Key | PTLRDCMBXHILCL-UHFFFAOYSA-M |
Synonym | sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c |
PubChem CID | 443495 |
ChEBI | CHEBI:29678 |
IUPAC Name | sodium;oxoarsinite |
SMILES | [O-][As]=O.[Na+] |
Description
Chemical Identifiers
7784-46-5 | |
129.909 | |
PTLRDCMBXHILCL-UHFFFAOYSA-M | |
443495 | |
sodium;oxoarsinite |
AsNaO2 | |
3472 | |
sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c | |
CHEBI:29678 | |
[O-][As]=O.[Na+] |
Specifications
7784-46-5 | |
Liquid | |
3472 | |
PTLRDCMBXHILCL-UHFFFAOYSA-M | |
sodium;oxoarsinite | |
443495 | |
HDPE plastic bottle |
Colorless | |
AsNaO2 | |
sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c | |
[O-][As]=O.[Na+] | |
129.909 | |
CHEBI:29678 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium arsenite solution 0.05M (0.1N)
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P281-Use personal protective equipment as required.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.