missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Sodium azide, 99+%, for biochemistry Product Code.: 12695107

Thermo Scientific Chemicals Sodium azide, 99+%, for biochemistry

Product Code. 12695107
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 12695107

Brand: Thermo Scientific Chemicals 447810250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium azide, CAS # 26628-22-8, is the sodium salt of hydrogen azide (hydrazoic acid). This compound shows antibacterial and mutagenic activities. It also can inhibit the mitochondrial respiratory chain.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Sodium azide is a precursor for preparing inorganic azides such as lead azide and silver azide
• One mechanism of action associated with this compound can be related to the inhibition of mitochondrial ATPase, that consequently suppresses oxidative phosphorylation

Applications

• Sodium azide is an inorganic compound with antibacterial and mutagenic activities mainly used in biological samples
• Sodium azide inhibits cytochrome oxidase in gram-negative bacteria. However, its antibacterial capacity is more limited in some gram-positive bacteria (streptococci, pneumococci, lactobacilli), presenting intrinsic resistance
• Sodium azide is used for the introduction of an azide group to prepare a desired compound
• It is useful in biochemical approaches as a probe reagent and a preservative
• This compound can be used to synthetize blue fluorescent copolymers and metal phosphonates

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.01
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid germanShow MoreShow Less
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Boiling Point 300.0°C
Melting Point 275.0°C
Quantity 25 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula NaN3
Merck Index 15,8717
Solubility Information Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether
Formula Weight 65.01
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic bottle
Specific Gravity 1.85
Density 1.8500g/mL
Chemical Name or Material Sodium azide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457282 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number447811000,447815000,447810250
Lot NumberA0445823 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number447811000,447815000,447810250
Lot NumberA0457282 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number447811000,447815000,447810250
Lot NumberA0382571 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number447811000,447815000,447810250
Lot NumberA0442130 Certificate TypeCertificate of Analysis Date20/05/2022 Catalog Number447811000,447815000,447810250
compliance-icons compliance-icons
Product Identifier
  • Sodium azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sodium azide, 99+%, for biochemistry >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.