missing translation for 'onlineSavingsMsg'
Learn More
Sodium azide, 99%, extra pure, Thermo Scientific Chemicals Product Code.: 10124813

Sodium azide, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10124813
2.5 kg
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 10124813

Brand: Thermo Scientific Acros 190380025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26628-22-8
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Boiling Point 300.0°C
Melting Point 275.0°C
Color White
pH 9 (5% min. soln.)
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98.5% min
Molecular Formula N3Na
Linear Formula NaN3
Fieser 01,1041; 02,376; 03,259; 04,440; 05,593; 15,289; 17,68
Merck Index 15, 8717
Solubility Information Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether
Molecular Weight (g/mol) 65.01
Formula Weight 65.01
Percent Purity 99%
Grade Extra Pure
Loss on Drying 0.5% max.
Packaging Plastic bottle
Specific Gravity 1.85
Decomposition Information >275°C
Density 1.8500g/mL
Chemical Name or Material Sodium azide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium azide, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.