missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, Certified AR for Analysis, Fisher Chemical™

Product Code. 10050520
Click to view available options
Quantity:
100 g
500 g

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number 3536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]

Specifications

Boiling Point 300°C
Melting Point 275°C
Color White
pH 10
Physical Form Powder/Solid
Quantity 100 g
Formula Weight 65.01g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium azide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P330-Rinse mouth.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'