missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Azide, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Boiling Point | 300°C |
Melting Point | 275°C |
Color | White |
pH | 10 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Formula Weight | 65.01g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Sodium azide
- Danger
- Acute toxicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H300-Fatal if swallowed.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'