missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Azide, White Powder, Fisher BioReagents
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle/Poison Pack
Description
- Sodium Azide is readily soluble in water
- Handle with caution and follow all storage recommendations
- For research use only
Specifications
Specifications
Boiling Point | 300°C |
Melting Point | 275°C |
CAS | 26628-22-8 |
Color | White |
pH | 10 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Molecular Formula | N3Na |
Merck Index | 15, 8717 |
Synonym | sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german |
Show More |
Safety and Handling
Product Identifier
- Sodium azide
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction