missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, 1M buffer soln., pH 8.5, Thermo Scientific Chemicals

Product Code. 15473179
Click to view available options
Quantity:
250 mL
500 mL

Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 8.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water
Chemical Name or Material Sodium bicarbonate buffer soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'