missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, 99+%, Extra Pure, Thermo Scientific Chemicals
Sodium bicarbonate, 99+%, CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 5kg, 20ppm max., 100ppm max., CHEBI:32139, White, 205-633-8
29.15 € - 223.11 €
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
SMILES | [Na+].OC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10694782
|
Thermo Scientific Acros
123360010 |
1 kg | Plastic bottle |
29.15 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10051210
|
Thermo Scientific Acros
123360050 |
5 kg | Plastic drum |
84.12 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10493635
|
Thermo Scientific Acros
123360250 |
25 kg | Fibre drum |
223.11 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
CHEBI:32139 |
Specifications
270.0°C | |
White | |
99+% | |
NaHCO3 | |
03,260; 05,595; 11,476; 17,180 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
84.01 | |
Extra Pure |
144-55-8 | |
Fine Crystalline Powder | |
CHNaO3 | |
MFCD00003528 | |
15, 8719 | |
Solubility in water: 9g/100mL (20°C). Other solubilities: 16.4g/100mL water (60°C),insoluble in alcohol | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
≥99% | |
Sodium bicarbonate |
Safety and Handling
missing translation for 'einecsNumber' : 205-633-8
Product Identifier
- Sodium bicarbonate
RUO – Research Use Only